Skip to Content
MilliporeSigma
Prostaglandin I2 sodium salt ≥96% (HPLC), synthetic, powder

Prostaglandin I2 sodium salt

Prostaglandin I2 sodium salt ≥96% (HPLC), synthetic, powder
Synonym(s):
(5Z,9α,11α,13E,15S)-6,9-Epoxy-11,15-dihydroxyprosta-5,13-dien-1-oic acid sodium salt, Epoprostenol sodium salt, PGI2-Na, Prostacyclin sodium salt
Empirical Formula (Hill Notation):
C20H31NaO5
CAS Number:
Molecular Weight:
374.45
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing