Skip to Content
MilliporeSigma
Propylene glycol propyl ether 99%

Propylene glycol propyl ether

Propylene glycol propyl ether 99%
Synonym(s):
1-Propoxy-2-propanol
Linear Formula:
CH3CH2CH2OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
118.17

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing