Skip to Content
MilliporeSigma
Propionitrile analytical standard

Propionitrile

Propionitrile analytical standard
Synonym(s):
Ethyl cyanide, Propionitrile
Linear Formula:
CH3CH2CN
CAS Number:
Molecular Weight:
55.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing