Skip to Content
MilliporeSigma
Proparacaine Hydrochloride pharmaceutical secondary standard, certified reference material

Proparacaine hydrochloride

Proparacaine Hydrochloride pharmaceutical secondary standard, certified reference material
Synonym(s):
Proxymetacaine hydrochloride, Proparacaine hydrochloride
Empirical Formula (Hill Notation):
C16H26N2O3 · HCl
CAS Number:
Molecular Weight:
330.85

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade