Skip to Content
MilliporeSigma
vibrant-m

Propafenone impurity B

vibrant-m
Synonym(s):
(2E)-1-{2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylprop-2-en-1-one
Empirical Formula (Hill Notation):
C21H25NO3
CAS Number:
Molecular Weight:
339.43

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing