Skip to Content
MilliporeSigma
Probucol Related Compound A pharmaceutical secondary standard, certified reference material

Probucol Related Compound A

Probucol Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
Probucol
Empirical Formula (Hill Notation):
C31H48O2S2
CAS Number:
Molecular Weight:
516.84

Products

Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1