Skip to Content
MilliporeSigma
Proadifen hydrochloride analytical standard, ≥95%

Proadifen hydrochloride

Proadifen hydrochloride analytical standard, ≥95%
Synonym(s):
α-Phenyl-α-propylbenzeneacetic acid 2-(diethylamino)ethyl ester, N,N-Diethylaminoethyl 2,2-diphenylvalerate, SKF-525A
Empirical Formula (Hill Notation):
C23H31NO2 · HCl
CAS Number:
Molecular Weight:
389.96

Products

Brand
Formula Weight
Gene Alias
Purity
Format
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing