Skip to Content
MilliporeSigma
vibrant-m

Prilocaine impurity E

vibrant-m
Synonym(s):
(RS)-N-(Methylphenyl)-2-(propylamino)propanamide
Empirical Formula (Hill Notation):
C13H20N2O
CAS Number:
Molecular Weight:
220.31

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing