Skip to Content
MilliporeSigma
Praseodymium nitrate hexahydrate 99.99% (trace rare earth metals basis)

Praseodymium nitrate hexahydrate

Praseodymium nitrate hexahydrate 99.99% (trace rare earth metals basis)
Synonym(s):
Praseodymium(III) nitrate hexahydrate, Praseodymium trinitrate hexahydrate
Linear Formula:
Pr(NO3)3·6H2O
CAS Number:
Molecular Weight:
435.01

Products

Product Category
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing