Skip to Content
MilliporeSigma
Potassium trimethylsilanolate 90%, technical grade

Potassium trimethylsilanolate

Potassium trimethylsilanolate 90%, technical grade
Synonym(s):
Trimethylsilanol potassium salt
Linear Formula:
(CH3)3SiOK
CAS Number:
Molecular Weight:
128.29
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing