Skip to Content
MilliporeSigma
Potassium trifluoromethanesulfonimide ≥95%

Potassium trifluoromethanesulfonimide

Potassium trifluoromethanesulfonimide ≥95%
Synonym(s):
Potassium bis(trifluoromethanesulfonyl)imide
Empirical Formula (Hill Notation):
C2F6KNO4S2
CAS Number:
Molecular Weight:
319.24

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing