Skip to Content
MilliporeSigma
Potassium tri(3,5-dimethyl-1-pyrazolyl)borohydride 97%

Potassium tri(3,5-dimethyl-1-pyrazolyl)borohydride

Potassium tri(3,5-dimethyl-1-pyrazolyl)borohydride 97%
Synonym(s):
Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate, Scorpionates, Tp ligand
Empirical Formula (Hill Notation):
C15H22BKN6
CAS Number:
Molecular Weight:
336.28

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing