Skip to Content
MilliporeSigma
Potassium thioacetate 98%

Potassium thioacetate

Potassium thioacetate 98%
Synonym(s):
Ethanethioic acid potassium salt, Thioacetic acid potassium salt, Thiolacetic acid potassium salt
Linear Formula:
CH3COSK
CAS Number:
Molecular Weight:
114.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing