Skip to Content
MilliporeSigma
Potassium tetrakis(4-chlorophenyl)borate Selectophore™, ≥98.0%

Potassium tetrakis(4-chlorophenyl)borate

Potassium tetrakis(4-chlorophenyl)borate Selectophore™, ≥98.0%
Synonym(s):
Tetrakis(4-chlorophenyl)boron potassium
Linear Formula:
(ClC6H4)4BK
CAS Number:
Molecular Weight:
496.11
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing