Skip to Content
MilliporeSigma
Potassium L-tartrate monobasic puriss., meets analytical specification of Ph.Eur., BP, FCC, 99.5-100.5% (related to dried substance)

Potassium L-tartrate monobasic

Potassium L-tartrate monobasic puriss., meets analytical specification of Ph.Eur., BP, FCC, 99.5-100.5% (related to dried substance)
Synonym(s):
L(+)-Tartaric acid monopotassium salt, Potassium bitartrate, Potassium hydrogen L-tartrate, Potassium hydrogen tartrate, Tartaric acid monopotassium salt, L-(+)-Tartaric acid monopotassium salt
Linear Formula:
KO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
188.18
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Special Grade