Skip to Content
MilliporeSigma
Potassium hexacyanocobaltate(III) ≥97.0%

Potassium hexacyanocobaltate(III)

Potassium hexacyanocobaltate(III) ≥97.0%
Synonym(s):
Tripotassium hexacyanocobaltate(III)
Linear Formula:
K3Co(CN)6
CAS Number:
Molecular Weight:
332.33
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing