Skip to Content
MilliporeSigma
Potassium cyanide-13C 99 atom % 13C

Potassium cyanide-13C

Potassium cyanide-13C 99 atom % 13C
Synonym(s):
13C Labeled potassium cyanide, 13C labeled KCN
Linear Formula:
K13CN
CAS Number:
Molecular Weight:
66.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing