Skip to Content
MilliporeSigma
Potassium 5-hydroxypentanoyltrifluoroborate

Potassium 5-hydroxypentanoyltrifluoroborate

Potassium 5-hydroxypentanoyltrifluoroborate
Empirical Formula (Hill Notation):
C5H9BF3KO2
CAS Number:
Molecular Weight:
208.03

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing