Skip to Content
MilliporeSigma
vibrant-m

Potassium 2-[(8-chloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctyl)oxy]-1,1,2,2-tetrafluoroethanesulfonate

vibrant-m
Synonym(s):
11-Chloroeicosafluoro-3-oxaundecane-1-sulfonic acid potassium salt, 8:2 Cl-PFAES
Empirical Formula (Hill Notation):
C10ClF20KO4S
CAS Number:
Molecular Weight:
670.69

Products

Brand
Formula Weight
Quality Segment
Reference Material Type