Skip to Content
MilliporeSigma
Posaconazole solution 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Posaconazole solution

Posaconazole solution 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C37H42F2N8O4
CAS Number:
Molecular Weight:
700.78
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique