Skip to Content
MilliporeSigma
Polystyrene thiol terminated average Mn 5,000, PDI ≤1.1

Polystyrene thiol terminated

Polystyrene thiol terminated average Mn 5,000, PDI ≤1.1
Synonym(s):
MercaptoPS, Mercaptopolystyrene, PS SH terminated
Linear Formula:
CNC3H6(C8H8)nSH

Products

Brand
Melting Point (°C)
Physical Form
Polymer Composition
Polymer Type
Showing 1-2 of 2