Skip to Content
MilliporeSigma
Polyethylene terephthalate G United States Pharmacopeia (USP) Reference Standard

Polyethylene terephthalate G

Polyethylene terephthalate G United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Poly(1,4-cyclohexanedimethylene terephthalate-co-ethylene terephthalate)
Linear Formula:
(-OCH2C6H10CH2O2CC6H4-4-CO-)x(-OCH2CH2O2CC6H4-4-CO-)y
CAS Number:

Products

Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1