Skip to Content
MilliporeSigma
Poly(ethylene glycol) methyl ether 2-bromoisobutyrate average Mn 600

Poly(ethylene glycol) methyl ether 2-bromoisobutyrate

Poly(ethylene glycol) methyl ether 2-bromoisobutyrate average Mn 600
Synonym(s):
ATRP initiator, PEG methyl ether bromoisobutyrate, mPEG bromoisobutyrate, PEG bib, mPEG ATRP, methoxyPolyethylene glycol bromoisobutyryl
Linear Formula:
CH3O(CH2CH2O)nCOC(CH3)2Br
CAS Number:

Products

Brand
Physical Form
Functional Group
Polymer Composition
Polymer Type