Skip to Content
MilliporeSigma
Poly(ethylene glycol) bis(3-aminopropyl) terminated Mn ~1,500

Poly(ethylene glycol) bis(3-aminopropyl) terminated

Poly(ethylene glycol) bis(3-aminopropyl) terminated Mn ~1,500
Synonym(s):
O,O′-Bis(3-aminopropyl)polyethylene glycol 1,500
Linear Formula:
(C2H4O)nC6H16N2O
CAS Number:

Products

Brand
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type