Skip to Content
MilliporeSigma
Polyethylene glycol 3000 monodisperse solution BioUltra, Molecular Biology, ~50% in H2O

Polyethylene glycol 3000 monodisperse solution

Polyethylene glycol 3000 monodisperse solution BioUltra, Molecular Biology, ~50% in H2O
Synonym(s):
Polyethylene glycol solution, PEG solution
CAS Number:

Products

Product Category
pH Value
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1