Poly(2-ethyl-2-oxazoline) 5-7 cSt
Synonym(s):
Poly(2-ethyl-2-oxazoline), PEOX, POx, PetOx, poly 2-ethyloxazoline, polyethyloxazoline
Linear Formula:
[-N(COC2H5)CH2CH2-]n
CAS Number:
Products
Brand
Physical Form
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing