Skip to Content
MilliporeSigma
Piroctone Related Compound B pharmaceutical secondary standard, certified reference material

Piroctone Related Compound B

Piroctone Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
4-Methyl-6-(2,4,4-trimethylpentyl)-2H-pyran-2-one
Empirical Formula (Hill Notation):
C14H22O2
CAS Number:
Molecular Weight:
222.32

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade