
Piperidine-1-sulfonyl chloride

Synonym(s):
1-Piperidinesulfonyl chloride
Empirical Formula (Hill Notation):
C5H10ClNO2S
CAS Number:
Molecular Weight:
183.66
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1