Skip to Content
MilliporeSigma
Piperazine-2-carboxylic acid dihydrochloride 98%

Piperazine-2-carboxylic acid dihydrochloride

Piperazine-2-carboxylic acid dihydrochloride 98%
Synonym(s):
(±)-Piperazine-2-carboxylic acid dihydrochloride
Empirical Formula (Hill Notation):
C5H10N2O2 · 2HCl
CAS Number:
Molecular Weight:
203.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing