![Picrylsulfonic acid solution 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines](/deepweb/assets/sigmaaldrich/product/structures/223/720/cbdbcfba-c731-4989-a636-437a6ee44585/640/cbdbcfba-c731-4989-a636-437a6ee44585.png)
Picrylsulfonic acid solution
![Picrylsulfonic acid solution 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines](/deepweb/assets/sigmaaldrich/product/structures/223/720/cbdbcfba-c731-4989-a636-437a6ee44585/640/cbdbcfba-c731-4989-a636-437a6ee44585.png)
Synonym(s):
2,4,6-Trinitrobenzenesulfonic acid solution, TNBS
Empirical Formula (Hill Notation):
C6H3N3O9S
CAS Number:
Molecular Weight:
293.17
Products
Product Category
Brand
Formula Weight
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing