Skip to Content
MilliporeSigma
Picric acid moistened with water, ≥98%

Picric acid

Picric acid moistened with water, ≥98%
Synonym(s):
2,4,6-Trinitrophenol, PA
Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing