Skip to Content
MilliporeSigma
Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt ≥98% (enzymatic)

Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt

Phospho(enol)pyruvic acid tri(cyclohexylammonium) salt ≥98% (enzymatic)
Synonym(s):
2-(Phosphonooxy)-2-propenoic acid tri(cyclohexylammonium) salt, PEP-3CHA, Phosphoenolpyruvic acid tris(cyclohexylamine) salt
Empirical Formula (Hill Notation):
C3H5O6P · 3C6H13N
CAS Number:
Molecular Weight:
465.56
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1