Skip to Content
MilliporeSigma
Phospho(enol)pyruvic acid monosodium salt hydrate ≥97% (enzymatic)

Phospho(enol)pyruvic acid monosodium salt hydrate

Phospho(enol)pyruvic acid monosodium salt hydrate ≥97% (enzymatic)
Synonym(s):
2-(Phosphonooxy)-2-propenoic acid monosodium, mono-Sodium phosphoenolpyruvate hydrate, Monosodium phosphoenolpyruvate
Empirical Formula (Hill Notation):
C3H4NaO6P · xH2O
CAS Number:
Molecular Weight:
190.02 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing