Skip to Content
MilliporeSigma
Phosphatidylinositol ammonium salt solution from soybean, ≥98.0% (TLC)

Phosphatidylinositol ammonium salt solution

Phosphatidylinositol ammonium salt solution from soybean, ≥98.0% (TLC)
Synonym(s):
L-α-Phosphatidylinositol ammonium salt, PI, PtdIns
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Chemical Composition
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing