Skip to Content
MilliporeSigma
Phenylmercury nitrate basic, ≥97.0% (RT)

Phenylmercury nitrate

Phenylmercury nitrate basic, ≥97.0% (RT)
Synonym(s):
Phenylmercuric nitrate, basic
Linear Formula:
C6H5HgNO3·C6H5HgOH
CAS Number:
Molecular Weight:
634.40

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing