Skip to Content
MilliporeSigma
Phenol Red sodium salt ACS reagent, Dye content 90 %

Phenol red sodium salt

Phenol Red sodium salt ACS reagent, Dye content 90 %
Synonym(s):
Phenol red sodium salt, Phenolsulfonephthalein sodium salt
Empirical Formula (Hill Notation):
C19H13NaO5S
CAS Number:
Molecular Weight:
376.36
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing