Skip to Content
MilliporeSigma
Phenolphthalein bisphosphate tetrasodium salt ~95%

Phenolphthalein bisphosphate tetrasodium salt

Phenolphthalein bisphosphate tetrasodium salt ~95%
Synonym(s):
Phenolphthalein diphosphate tetrasodium salt
Linear Formula:
C20H12O10P2Na4
CAS Number:
Molecular Weight:
566.21

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing