Skip to Content
MilliporeSigma
Phenanthrene-d10 98 atom % D, 98% (CP)

Phenanthrene-d10

Phenanthrene-d10 98 atom % D, 98% (CP)
Empirical Formula (Hill Notation):
C14D10
CAS Number:
Molecular Weight:
188.29

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing