Skip to Content
MilliporeSigma
Perfluoroundecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Perfluoroundecanoic acid

Perfluoroundecanoic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Heneicosafluoroundecanoic acid, PFUnDA
Linear Formula:
CF3(CF2)9CO2H
CAS Number:
Molecular Weight:
564.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade