Penicillin V potassium
Synonym(s):
Penicillin V potassium salt, Phenoxymethylpenicillin potassium salt
Linear Formula:
C16H17N2O5SK
CAS Number:
Molecular Weight:
388.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2