Skip to Content
MilliporeSigma
Paracetamol sulfate potassium salt solid, ≥97% (HPLC)

Paracetamol sulfate potassium salt

Paracetamol sulfate potassium salt solid, ≥97% (HPLC)
Synonym(s):
4-Acetamidophenol sulfate ester potassium salt, Acetaminophen sulfate potassium salt, N-(4-Sulfoxyphenyl)acetamide monopotassium salt
Empirical Formula (Hill Notation):
C8H8KNO5S
CAS Number:
Molecular Weight:
269.32

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing