Skip to Content
MilliporeSigma
Pantoprazole Related Compound B pharmaceutical secondary standard, certified reference material

Pantoprazole Related Compound B

Pantoprazole Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
5-(Difluoromethoxy)-2-{[(3,4-dimethoxy-2-pyridinyl)methyl]thio}-1H-benzimidazole
Empirical Formula (Hill Notation):
C16H15F2N3O3S
CAS Number:
Molecular Weight:
367.37

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade