Skip to Content
MilliporeSigma
Palonosetron hydrochloride ≥98% (HPLC)

Palonosetron Hydrochloride

Palonosetron hydrochloride ≥98% (HPLC)
Synonym(s):
Palonosetron hydrochloride, (3aS)-2-[(3S)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride, (3aS)-2-[(3S)-1-Azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-1H-Benz[de]isoquinolin-1-one hydrochloride (1:1), RS 25259-197
Empirical Formula (Hill Notation):
C19H24N2O · HCl
CAS Number:
Molecular Weight:
332.87

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade