Skip to Content
MilliporeSigma
Palmitoyl chloride 98%

Palmitoyl chloride

Palmitoyl chloride 98%
Synonym(s):
Hexadecanoyl chloride, Palmitoyl chloride
Linear Formula:
CH3(CH2)14COCl
CAS Number:
Molecular Weight:
274.87

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing