Skip to Content
MilliporeSigma
Palladium(II) nitrate dihydrate (40% Pd) for synthesis

Palladium(II) nitrate dihydrate

Palladium(II) nitrate dihydrate (40% Pd) for synthesis
Synonym(s):
Palladium(II) nitrate dihydrate (40% Pd)
Linear Formula:
Pd(NO3)2 · 2 H2O
CAS Number:
Molecular Weight:
266.46

Products

Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing