Skip to Content
MilliporeSigma
P1,P5-Di(adenosine-5′) pentaphosphate trilithium salt ≥95%, powder

P1,P5-Di(adenosine-5′) pentaphosphate trilithium salt

P1,P5-Di(adenosine-5′) pentaphosphate trilithium salt ≥95%, powder
Synonym(s):
A(5′)P5(5′)A trilithium salt, Diadenosine pentaphosphate trilithium salt
Empirical Formula (Hill Notation):
C20H26Li3N10O22P5
CAS Number:
Molecular Weight:
934.17
EC Number:

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing