Skip to Content
MilliporeSigma
Oxyfluorfen PESTANAL®, analytical standard

Oxyfluorfen

Oxyfluorfen PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing