Skip to Content
MilliporeSigma
Oxalyl bromide 97%

Oxalyl bromide

Oxalyl bromide 97%
Synonym(s):
Ethanedioyl dibromide, NSC 96957
Linear Formula:
BrCOCOBr
CAS Number:
Molecular Weight:
215.83
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing