Skip to Content
MilliporeSigma
O-Tritylhydroxylamine 95%

O-Tritylhydroxylamine

O-Tritylhydroxylamine 95%
Synonym(s):
O-(Triphenylmethyl)hydroxylamine, Trityloxyamine
Linear Formula:
(C6H5)3CONH2
CAS Number:
Molecular Weight:
275.34
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing