Skip to Content
MilliporeSigma
Orlistat Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material

Orlistat Related Compound B

Orlistat Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Diisopropyl hydrazine-1,2-dicarboxylate, Diisopropyl bicarbamate
Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade